missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-9-phenylfluorene, 97%

Product Code. 10361290 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10361290 1 g Glass bottle 1g
10851671 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10361290 Leverandør Thermo Scientific Acros Leverandørnr. 428610010
 Restricted Item
Estimated Shipment: 29-09-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 55135-66-5 | C19H13Br | 321.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 55135-66-5
Molecular Formula C19H13Br
Molecular Weight (g/mol) 321.21
MDL Number MFCD00075522
InChI Key HYQXNCDBSALQLB-UHFFFAOYSA-N
Synonym 9-bromo-9-phenyl-9h-fluorene, 9-brom-9-phenyl-9h-fluoren, zlchem 855, acmc-209ll7, 9-bromo-9-phenyl-fluorene, 9-bromanyl-9-phenyl-fluorene, hyqxncdbsalqlb-uhfffaoysa, 9-bromo-9-phenylfluorene, 9h-fluorene, 9-bromo-9-phenyl
PubChem CID 231624
IUPAC Name 9-bromo-9-phenylfluorene
SMILES C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)Br

Tekniske data

CAS Min % 96.0
CAS Max % 100.0
Melting Point 99°C to 101°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 321.21
Percent Purity 97%
Chemical Name or Material 9-Bromo-9-phenylfluorene
compliance-icons
Produktidentifikator
  • 9-Bromo-9-phenylfluorene
Signalord
  • Danger
Farekategori
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Faresætning
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.