missing translation for 'onlineSavingsMsg'
Learn More

9-Bromoanthracene, TRC

Product Code. 30180370
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180370 5 g 5g
30180060 25 g 25g
30180286 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180370 Supplier Toronto Research Chemicals Supplier No. TRCB6798005G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1564-64-3
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
Synonym 10-Bromoanthracene, 9-Anthracenyl bromide, 9-Anthracyl bromide, NSC 803
IUPAC Name 9-bromoanthracene
SMILES Brc1c2ccccc2cc3ccccc13

Specifications

Quantity 5 g
InChI Formula InChI=1S/C14H9Br/c15-14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-9H
Formula Weight 255.9888
Chemical Name or Material 9-Bromoanthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.