missing translation for 'onlineSavingsMsg'
Learn More

9-Chloroanthracene (>90%), TRC

Product Code. 30362942
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30362942 1 g 1g
30358171 250 mg 250mg
30353043 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30362942 Supplier Toronto Research Chemicals Supplier No. TRCC3644031G
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 716-53-0
Molecular Formula C14 H9 Cl
Molecular Weight (g/mol) 212.67
Synonym 9-Chloroanthracene (ACI), 10-Chloroanthracene, 9-Chloroanthracene (ACI)
IUPAC Name 9-chloroanthracene
SMILES Clc1c2ccccc2cc3ccccc13

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H9Cl/c15-14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-9H
Formula Weight 212.039
Chemical Name or Material 9-Chloroanthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.