missing translation for 'onlineSavingsMsg'
Learn More

9-Chloroanthracene, 96%, Thermo Scientific Chemicals

Product Code. 11324454
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324454 5 g 5g
11334454 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324454 Supplier Thermo Scientific Alfa Aesar Supplier No. L01092.06
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 716-53-0 | Molecular Formula: C14H9Cl | Molecular Weight: 212.676 g/mol

9-Chloroanthracene, is used as a chlorinated anthracene compound used for biochemical research. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Chloroanthracene, is used as a chlorinated anthracene compound used for biochemical research. It is also used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 716-53-0
Molecular Formula C14H9Cl
Molecular Weight (g/mol) 212.676
MDL Number MFCD00001245
InChI Key KULLJOPUZUWTMF-UHFFFAOYSA-N
PubChem CID 69744
IUPAC Name 9-chloroanthracene
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2Cl

Specifications

Melting Point 101°C to 106°C
Quantity 5 g
Beilstein 1869540
Synonym anthracene, 9-chloro, ccris 5547, 9-chloro-anthracene, #, anthracene,9-chloro, bidd:gt0730
Solubility Information Insoluble in water.
Formula Weight 212.68
Percent Purity 96%
Chemical Name or Material 9-Chloroanthracene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.