missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenemethanol, 99%

Product Code. 10285840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10285840 1 g Glass bottle 1g
10539073 5 g Glass bottle 5g
10439460 10 g Glass bottle 10g
10731935 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10285840 Supplier Thermo Scientific Acros Supplier No. 170930010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 24324-17-2 | C14H12O | 196.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24324-17-2
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00001139
InChI Key XXSCONYSQQLHTH-UHFFFAOYSA-N
Synonym 9-fluorenemethanol, 9-fluorenylmethanol, 9h-fluorene-9-methanol, 9-fluorenyl methanol, 9h-fluoren-9-yl methanol, fluorene-9-methanol, 9-hydroxymethylfluorene, 9-fluorenmethanol, hofm, fluoren-9-yl methanol
PubChem CID 90466
SMILES OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 104°C to 107°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Beilstein 06, III, 3507
Fieser 05,308; 03,145; 04,237; 13,134
IUPAC Name (9H-fluoren-9-yl)methanol
Formula Weight 196.25
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenemethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.