missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone hydrazone, 97%, Thermo Scientific™

Product Code. 15214614 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15214614 5 g Glass bottle 5g
15224614 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15214614 Supplier Thermo Scientific Acros Supplier No. 119230050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13629-22-6
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.24
InChI Key YCNUILAKOMIBAL-UHFFFAOYSA-N
Synonym 9-fluorenone hydrazone, fluoren-9-one, hydrazone, fluorenone hydrazone, 9h-fluoren-9-ylidene hydrazine, 9h-fluoren-9-one, hydrazone, fluoren-9-one hydrazone, 9h-fluoren-9-ylidenehydrazine, fluorenonhydrazon, fluoren-9-ylidenediazane, fluoren-9-ylidene-hydrazine
PubChem CID 83627
IUPAC Name fluoren-9-ylidenehydrazine
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2=NN

Specifications

Melting Point 148.0°C to 150.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 07,I,251
Solubility Information Solubility in water: insoluble
Formula Weight 194.24
Percent Purity 97%
Chemical Name or Material 9-Fluorenone hydrazone
compliance-icons
Product Identifier
  • 9-Fluorenone hydrazone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.