missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone hydrazone, 97%

Product Code. p-7025960
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420307 5 g 5g
11430307 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420307 Supplier Thermo Scientific Alfa Aesar Supplier No. B21706.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13629-22-6 | C13H10N2 | 194.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13629-22-6
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.237
MDL Number MFCD00016357
InChI Key YCNUILAKOMIBAL-UHFFFAOYSA-N
Synonym 9-fluorenone hydrazone, fluoren-9-one, hydrazone, fluorenone hydrazone, 9h-fluoren-9-ylidene hydrazine, 9h-fluoren-9-one, hydrazone, fluoren-9-one hydrazone, 9h-fluoren-9-ylidenehydrazine, fluorenonhydrazon, fluoren-9-ylidenediazane, fluoren-9-ylidene-hydrazine
PubChem CID 83627
IUPAC Name fluoren-9-ylidenehydrazine
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2=NN

Specifications

Melting Point 150°C to 154°C
Quantity 5 g
Beilstein 1910575
Formula Weight 194.24
Percent Purity ≥98%
Chemical Name or Material 9-Fluorenone hydrazone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.