missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone oxime, 98+%, Thermo Scientific Chemicals

Product Code. 11346726
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11346726 5 g 5g
11356726 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346726 Supplier Thermo Scientific Alfa Aesar Supplier No. L03950.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2157-52-0 | Molecular Formula: C13H9NO | Molecular Weight: 195.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2157-52-0
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00016356
InChI Key CRNNFEKVPRFZKJ-UHFFFAOYSA-N
Synonym 9-fluorenone oxime, 9h-fluoren-9-one oxime, fluorenone oxime, 9-oximinofluorene, fluorenone-9-oxime, 9h-fluoren-9-one, oxime, fluoren-9-one, oxime, fluoren-9-one oxime, 9-hydroxyimino fluorene, fluorenonoxim
PubChem CID 16543
IUPAC Name N-fluoren-9-ylidenehydroxylamine
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2=NO

Specifications

Melting Point 193°C to 194°C
Quantity 5 g
Beilstein 1871046
Formula Weight 195.22
Percent Purity ≥98%
Chemical Name or Material 9-Fluorenone oxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.