missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenylmethanol, 99%

Product Code. 11492228
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492228 5 g 5g
11402238 25 g 25g
11412238 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492228 Supplier Thermo Scientific Alfa Aesar Supplier No. A15212.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 24324-17-2 | C14H12O | 196.25 g/mol

9-Fluorenylmethanol acts as a N-protecting reagent, which is used in the synthesis of peptide. It is also used in the preparation of deoxynucleoside 9-fluorenemethyl phosphorodithioates. Further, it is used to prepare 9-(fluoromethyl)fluorene. In addition to this, it is involved in the electropolymerization with boron trifluoride diethyl etherate to yield low-potential electrodeposition of semiconducting poly(9-fluorenemethanol) film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Fluorenylmethanol acts as a N-protecting reagent, which is used in the synthesis of peptide. It is also used in the preparation of deoxynucleoside 9-fluorenemethyl phosphorodithioates. Further, it is used to prepare 9-(fluoromethyl)fluorene. In addition to this, it is involved in the electropolymerization with boron trifluoride diethyl etherate to yield low-potential electrodeposition of semiconducting poly(9-fluorenemethanol) film.

Solubility
Soluble in methanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24324-17-2
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00001139
InChI Key XXSCONYSQQLHTH-UHFFFAOYSA-N
Synonym 9-fluorenemethanol, 9-fluorenylmethanol, 9h-fluorene-9-methanol, 9-fluorenyl methanol, 9h-fluoren-9-yl methanol, fluorene-9-methanol, 9-hydroxymethylfluorene, 9-fluorenmethanol, hofm, fluoren-9-yl methanol
PubChem CID 90466
SMILES OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 102°C to 106°C
Quantity 5 g
Beilstein 2330017
Solubility Information Soluble in methanol.
IUPAC Name (9H-fluoren-9-yl)methanol
Formula Weight 196.25
Percent Purity 99%
Chemical Name or Material 9-Fluorenylmethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.