missing translation for 'onlineSavingsMsg'
Learn More

9-Nitro Minocycline Sulfate Salt (85%), TRC

Product Code. 30134718
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134718 100 mg 100mg
30003778 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134718 Supplier Toronto Research Chemicals Supplier No. TRCN494015100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153621-80-8
Molecular Formula C23H26N4O9 . 2(H 2SO4)
Molecular Weight (g/mol) 698.62
Synonym (4S,4aS,5aR,12aS)-4,7-Bis(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,10,12,12a-tetrahydroxy-9-nitro-1,11-dioxo-2-naphthacenecarboxamide Sulfate Salt, [4S-(4α,4aα,5aα,12aα)]-4,7-Bis(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,10,12,12a-tetrahydroxy-9-nitro-1,11-dioxo-2-naphthacenecarboxamid Sulfate Salt ;
IUPAC Name (4S,4aS,5aR,12aS)-4,7-bis(dimethylamino)-3,10,12,12a-tetrahydroxy-9-nitro-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide bis(sulfate)
SMILES O=S(O)(O)=O.O[C@]12[C@](C[C@@]3([H])C(C(C4=C(O)C(N(=O)=O)=CC(N(C)C)=C4C3)=O)=C2O)([H])[C@@H](C(O)=C(C1=O)C(N)=O)N(C)C.O=S(O)(O)=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C23H26N4O9.2H2O4S/c1-25(2)11-7-12(27(35)36)17(28)14-9(11)5-8-6-10-16(26(3)4)19(30)15(22(24)33)21(32)23(10,34)20(31)13(8)18(14)29;2*1-5(2,3)4/h7-8,10,16,28,30-31,34H,5-6H2,1-4H3,(H2,24,33);2*(H2,1,2,3,4)/t8-,10-,16-,23-;;/m0../s1
Formula Weight 698.11
Chemical Name or Material 9-Nitrominocycline Disulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.