missing translation for 'onlineSavingsMsg'
Learn More

9-Phenanthrenecarboxaldehyde, Thermo Scientific Chemicals

Product Code. 10157553 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10157553 5 g Glass bottle 5g
10147743 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10157553 Supplier Thermo Scientific Acros Supplier No. 130110050
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4707-71-5
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.24
MDL Number MFCD00001175
InChI Key QECIGCMPORCORE-UHFFFAOYSA-N
Synonym 9-phenanthrenecarboxaldehyde, phenanthrene-9-carboxaldehyde, 9-formylphenanthrene, phenanthrene-9-aldehyde, 9-phenanthrenecarbaldehyde, 9-phenanthrenealdehyde, 9-phenanthrencarbaldehyd, 9-phenanthraldehyde, 9-phenanthrenaldehyde, phenanthren-9-aldehyd
PubChem CID 78437
IUPAC Name phenanthrene-9-carbaldehyde
SMILES O=CC1=CC2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 100°C
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Formula Weight 206.24
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 9-Phenanthrenecarboxaldehyde
compliance-icons
Product Identifier
  • 9-Phenanthrenecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.