accessibility menu, dialog, popup

UserName

O

CAS RN 4707-71-5

IUPAC Name: phenanthrene-9-carbaldehyde
Synonyms: phenanthrene-9-carbaldehyde
Molecular Weight (g/mol): 206.24
Molecular Formula: C15H10O
InChi Key: QECIGCMPORCORE-UHFFFAOYSA-N
SMILES: O=CC1=CC2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results