missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate

Product Code. 10517742 Shop All Corning Products
Change view
Click to view available options
Packaging:
Bag of 20; Cs. of 100
Individually Wrapped
Unit Size:
Pack of 100
Pack of 48
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Unit Size
10517742 Individually Wrapped Pack of 48
10601442 Bag of 20; Cs. of 100 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10517742 Supplier Corning™ Supplier No. 3610
  • ONLINE EXCLUSIVE PRICE
    685.93 €
    610.94 € / Pack of 48
    Save 74.99 €
    10% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Color: White, Clear Bottom; Sterility: Sterile; Lid: With Lid

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Barcode No Barcode
    Format Standard
    Lid With Lid
    Culture Area 0.32 cm2
    Array 8 x 12
    Packaging Individually Wrapped
    Color White, Clear Bottom
    Product Type Microplate
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Sterile
    Surface Treatment Cell Culture
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Attachment and growth of anchorage-dependent cells
    No. per Case 48/Cs.
    No. per Pack 1/Pk.
    Quantity 48/Cs.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.