missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Product Code. p-9045031 Shop All Corning Products
Click to view available options
Packaging:
Bag of 20; Cs. of 100
Individually Wrapped
Unit Size:
Pack of 100
Pack of 48
This item is not returnable. View return policy

Product Code. 10517742

Brand: Corning™ 3610

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Color: White, Clear Bottom; Sterility: Sterile; Lid: With Lid

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Standard
    Lid With Lid
    Culture Area 0.32 cm2
    Array 8 x 12
    Packaging Individually Wrapped
    Color White, Clear Bottom
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Sterile
    Surface Treatment Cell Culture
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Attachment and growth of anchorage-dependent cells
    No. per Case 48/Cs.
    No. per Pack 1/Pk.
    Quantity 48/Cs.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.