missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 96-Well Flat Bottom PCR Microplates
GREENER_CHOICE

Product Code. p-8003382 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Gray
White, Black
White, Clear Bottom
Quantity:
240/Pk.
80/Pk.
Unit Size:
Pack of 240
Pack of 80
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity unitSize
15204526 White, Black 80/Pk. Pack of 80
10713224 White, Clear Bottom 80/Pk. Pack of 80
10298393 White, Clear Bottom 240/Pk. Pack of 240
15294516 Gray 80/Pk. Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15204526 Supplier Eppendorf™ Supplier No. 0030601700
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For quantitative protein analysis

TRUSTED_SUSTAINABILITY

Specifications

Certified Yes
Format Standard
Detection Method Quantitative Protein Analysis
Array 8 x 12
Packaging 5 Bags x 16 Plates
Color White, Black
Material Polypropylene
No. of Wells 96
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) For PCR techniques
Max. Heat Stability 121°C
Quantity 80/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.