missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-Well Microplate CELLSTAR™, Cell Culture-Treated
GREENER_CHOICE

Product Code. 10369081
Change view
Click to view available options
Barcode:
Includes Barcode
No Barcode
Unit Size:
Pack of 32
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Barcode Unit Size
10369081 No Barcode Pack of 32
32043092 Includes Barcode Pack of 32
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10369081 Supplier Greiner Bio-One Supplier No. 655090
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for cell culture and microscopy using fluorescence or luminescence detection. Optimized for low to medium magnification.

  • ANSI-compliant footprint for compatibility with automated systems
  • Physical surface treatment for improved cell adhesion
  • Enhanced cell attachment and higher proliferation rates to improve and accelerate cell expansion
  • Free of detectable DNase, RNase, and human DNA
  • Non-pyrogenic
  • Non-cytotoxic
  • High optical clarity for optimal microscopic examination
  • Stackable design for efficient storage and handling
  • Chimney well design allows each individual well to stand on its own, minimizing the risk of sample carryover and cross-contamination
  • Manufactured under DIN ISO 9001 guidelines
  • Supplied in a pack of 32
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance ANSI (ANSI/SBS 1-2004)
Format Standard
Lid With Lid
Culture Area 34 mm2
Array 8 x 12
Color Black, Clear Bottom
For Use With (Equipment) Automated systems
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 392 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) For research use
Quantity 32/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.