Learn More
Fisherbrand™ 96-Well Microplate Holder
Product Code. 15544120 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
54 x 19 x 19 mm
128 x 86 x 23 mm
Unit Size:
Each
Product Code. Dimensions (L x W x H) unitSize
15544120 128 x 86 x 23 mm Each
15865911 54 x 19 x 19 mm Each
2 options
Product Code. 15544120
Only null left
Add to basket
Add to basket
For use with Fisherbrand™ Microplate Shaker and Incubating Microplate Shaker
TRUSTED_SUSTAINABILITY
Specifications
Product Type 96-well Microplate Holder
Dimensions (L x W x H) 128 x 86 x 23 mm