Learn More
Greiner Bio-One 96-well Non Binding Microplates
GREENER_CHOICE
Product Code. 15901661
Change view
Click to view available options
Well Shape:
Round Bottom (U-well)
Unit Size:
Pack of 40
Product Code. Well Shape unitSize
15901661 Round Bottom (U-well) Pack of 40
1 options
Product Code. 15901661 Supplier Greiner Bio-One Supplier No. 650901
Add to basket
Add to basket
Features long-term surface stability without degradation or leaching.
  • Ultra low non-specific biomolecular binding properties (proteins, DNA, RNA)
  • Long-term surface stability without degradation or leaching
  • Higher assay sensitivity with reduced background
  • Significantly increase assay sensitivity
TRUSTED_SUSTAINABILITY
Specifications
Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Binding Type Non Binding
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Protein-Repellent
Total Volume (Metric) Well 323 μL
Well Shape Round Bottom (U-well)
Max. RCF 1000 x g (swinging-bucket rotor)
No. per Case 40
Quantity 40/Cs.
Well Color Clear
Well Design Chimney