Learn More
Greiner Bio-One 96-well Polypropylene Conical Bottomed Microplates
GREENER_CHOICE
Product Code. 13499826
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
Product Code. Color Unit Size
13499826 Clear Pack of 100
1 options
Product Code. 13499826 Supplier Greiner Bio-One Supplier No. 651261
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
For high-throughput screening and assay
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Ideally suited for agglutination tests
  • No sharp corners, which means that they can be pipetted easily and cleanly
  • Suitable for +/- analyses
  • Solid bottom
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Non Binding
Color Clear
Product Type Storage Microplate
Material Polypropylene
Volume (Metric) Well 50 to 335 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Untreated
Total Volume (Metric) Well 340 μL
Well Shape Round, Conical Bottom (V-well)
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 100
No. per Pack 10
Quantity 100/Cs.
Well Color Clear
Well Design Chimney