missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ 96-Well Polystyrene Micro Test Plates

Product Code. 11315872
Click to view available options
Well Shape:
Conical Bottom (V-well)
Round Bottom (U-well)
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 11315872

Brand: Ratiolab™ 6018111

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

With a precise design, perfectly shaped for use with automatic systems.

  • Manufactured from clear polystyrene
  • 96 well with volume approx. 300 μL
  • With U- or V-bottom
  • SBS standard micro test plate format
  • 8 x 12 wells alphanumerically coded
  • Wells with raised rims for reduced cross-contamination and effective sealing with Ratiolab cover films
  • Optimum space for precise positioning
  • Space-saving and stackable
  • Provided in packs of 100

Specifications

Barcode Alphanumeric
Format SBS Standard
Array 8 x 12
Color Clear
For Use With (Equipment) Automatic systems
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Well Shape Round Bottom (U-well)
Quantity 100/Pk.
Rimmed Raised
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.