missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
BIOPHARMAGREENER_CHOICE

Product Code. 10213073
Change view
Click to view available options
Temperature Range (Metric):
-20°C to +120°C
-20°C to +130°C
-20°C to +55°C
Well Shape:
Round
Square
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Well Shape Unit Size
10213073 -20°C to +130°C Square Pack of 50
10354033 -20°C to +130°C Round Pack of 50
10293223 -20°C to +55°C Round Pack of 50
10722684 -20°C to +120°C Round Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10213073 Supplier Thermo Scientific™ Supplier No. AB0675
  • ONLINE EXCLUSIVE PRICE
    190.00 €
    173.00 € / Pack of 50
    Save 17.00 €
    8% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Round or square CapMats, Sealing Mats, MatCaps for sealing 96-well storage plates, available in autoclavable and pierceable formats.

Thermo Scientific 96-well Sealing Mats are designed for 96-well storage plates and are available in either pierceable or autoclavable formats. The round or square capped mats are made of thermoplastic elastomer and are suitable for long-term storage.

Highlights

  • Certified RNAse, DNAse, PCR Inhibitor, and human DNA free
  • Round or square capped mats for sealing 96-well storage plates
  • Alphanumeric grid-referencing
  • Molded from a proprietary thermoplastic elastomer for a flexible design
  • Suitable for long term storage
  • Available in autoclavable or pierceable formats

Pierceable version:

  • Solvent resistant (including DMSO, Ethanol, and Isopropanol)
  • Suitable for Gamma Irradiation
  • Requires 6N for piercing
  • Temperature Range: -20°C to +55°C

Autoclavable version:

  • Suitable for Gamma Irradiation
  • Temperature Range: -20°C to +130°C

Hazardous: No

Storage Condition: Ambient

TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Solvent Or Alcohol Resistant Alcohol Resistant, Solvent Resistant
Description CapMat for 96-well Polypropylene Sample Processing & Storage Plates
Certifications/Compliance Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Material TPE
Sterility Sterile
Autoclavable Autoclavable
No. of Wells 96
No. per Case 50
No. per Pack 10
Plug Diameter 8.4mm
Temperature Range (English) -4°F to 266°F
For Use With (Application) Designed for sealing 96-well storage plate and suitable for long term storage
Format 96-well
Product Type 96-well capmat, sealing mat
Product Line Abgene
Adhesive None
Certified RNase/DNase-free Yes
Pierceable No
Temperature Range (Metric) -20°C to +130°C
Well Shape Square
Plate Compatibility Abgene 96 Square Well
Fit for Purpose PCR Ready
Green Features ACT Ecolabel (Silver)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.