missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ PCR Plates 96-well, 150 μL - PCR Plates
GREENER_CHOICE

Product Code. 10619779 Shop All Eppendorf Products
Click to view available options
Color:
Blue
Colorless
Skirt Style:
Skirted
Unskirted
Unskirted, Divisible
Product Type:
Divisible PCR Plates
Forensic DNA Grade PCR Plates
Low Profile PCR Plates
Microbiology PCR Plates
Packaging:
5 Bags x 4 Plates
Individually Blister Wrapped
Unit Size:
Pack of 10
Pack of 20
This item is not returnable. View return policy

Product Code. 10619779

Brand: Eppendorf™ 0030133307

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. Raised well rims enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.

  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.

Certifications

Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.

TRUSTED_SUSTAINABILITY

Spécification

Barcode No Barcode
Certifications/Compliance Human DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 4 Plates
Color Colorless
Product Type Low Profile PCR Plates
Skirt Style Unskirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 150 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) Ideal for extreme applications such as multiple heat sealing.
Max. RCF 2250 x g
Profile Low Profile
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 20 Pk.
Well Color Colorless
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis