missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 96-Well twin.tec™ Skirted PCR Plates
GREENER_CHOICE

Product Code. 10360485 Shop All Eppendorf Products
Click to view available options
Color:
Clear
Yellow, Clear
Unit Size:
Pack of 300
This item is not returnable. View return policy

Product Code. 10360485

Brand: Eppendorf™ 0030128990

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certified Yes
Format Standard
Detection Method Quantitative Protein Analysis
Array 8 x 12
Color Yellow, Clear
Product Type 96-well PCR Plate
Skirt Style Skirted
Material Polycarbonate
No. of Wells 96
Dnase Free Yes
For Use With (Application) For PCR techniques
Max. Heat Stability 121°C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.