missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 96-Well twin.tec™ Skirted PCR Plates
GREENER_CHOICE

Product Code. 10714977 Shop All Eppendorf Products
Click to view available options
Color:
Clear
Yellow, Clear
Unit Size:
Pack of 300
This item is not returnable. View return policy

Product Code. 10714977

Brand: Eppendorf™ 0030128770

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Certified Yes
Format Standard
Detection Method Quantitative Protein Analysis
Array 8 x 12
Color Clear
Material Polycarbonate
No. of Wells 96
Dnase Free Yes
For Use With (Application) For PCR techniques
Max. Heat Stability 121°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.