missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ 96-Well twin.tec™ Skirted PCR Plates
Shop All Eppendorf Products

Click to view available options
Color:
Clear
Yellow, Clear
Unit Size:
Pack of 300
Specifications
Specifications
| Certified | Yes |
| Format | Standard |
| Detection Method | Quantitative Protein Analysis |
| Array | 8 x 12 |
| Color | Clear |
| Material | Polycarbonate |
| No. of Wells | 96 |
| Dnase Free | Yes |
| For Use With (Application) | For PCR techniques |
| Max. Heat Stability | 121°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction