missing translation for 'onlineSavingsMsg'
Learn More

9H-Fluoren-9-ylmethyl 4-(3-butenoyl)tetrahydro-1(2h)-pyrazinecarboxylate, ≥97%, Thermo Scientific™

Product Code. 10356614
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10356614 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10356614 Supplier Thermo Scientific Maybridge Supplier No. MO07782DA
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 876728-43-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C23H24N2O3
MDL Number MFCD08271967
Quantity 1 g
Synonym 9h-fluoren-9-yl methyl 4-but-3-enoyl piperazine-1-carboxylate, 9h-fluoren-9-ylmethyl 4-3-butenoyl tetrahydro-1 2h-pyrazinecarboxylate, 9h-fluoren-9-ylmethyl 4-but-3-enoyl piperazine-1-carboxylate, 1-piperazinecarboxylicacid, 4-1-oxo-3-buten-1-yl-, 9h-fluoren-9-ylmethyl ester, 9h-fluoren-9-ylmethyl-4-3-butenoyl tetrahydro-1 2h-pyrazinecarboxylate
InChI Key WRSRYPVHNSMIQR-UHFFFAOYSA-N
SMILES C=CCC(=O)N1CCN(CC1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
IUPAC Name 9H-fluoren-9-ylmethyl 4-but-3-enoylpiperazine-1-carboxylate
Molecular Weight (g/mol) 376.456
PubChem CID 18525938
Formula Weight 376.45
Percent Purity 97%
Chemical Name or Material 9H-Fluoren-9-ylmethyl 4-(3-butenoyl)tetrahydro-1(2H)-pyrazinecarboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.