missing translation for 'onlineSavingsMsg'
Learn More

(9Z,11E)-Prodlure, MedChemExpress

Product Code. 16463538
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16463538

Brand: Medchem Express HY101735

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress (9Z,​11E)​-​Prodlure ((9Z,11E)-Tetradecadien-1-yl acetate) is the main component of the sex pheromone of female Spodoptera littoralis.

(9Z,​11E)​-​Prodlure ((9Z,11E)-Tetradecadien-1-yl acetate) is the main component of the sex pheromone of female Spodoptera littoralis.

Specifications

Chemical Name or Material (9Z,11E)-Prodlure
Physical Form Liquid
CAS 50767-79-8
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H28O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym (9Z,11E)-Tetradecadien-1-yl acetate Ferodin SL Litlure A
Solubility Information DMSO : 100 mg/mL (396.21 mM; Need ultrasonic)
SMILES CC/C=C/C=C\CCCCCCCCOC(C)=O
Molecular Weight (g/mol) 252.39
Formula Weight 252.39
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.