missing translation for 'onlineSavingsMsg'
Learn More

A 839977, MedChemExpress

Product Code. 16495928
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495928 10 mg 10mg
16482077 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16495928 Supplier Medchem Express Supplier No. HY13954
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress A 839977 is a P2X7 selective antagonist; it blocks BzATP-evoked calcium influx at recombinant human, rat and mouse P2X7 receptors (IC50 values are 20 nM, 42 nM and 150 nM respectively) and reduces inflammatory and neuropathic pain in animal models; the antihyperalgesic effects of P2X7 receptor blockade are mediated by blocking the release of IL-1beta.

A 839977 is a P2X7 selective antagonist

Specifications

Chemical Name or Material A 839977
CAS 870061-27-1
Color White
Purity Grade Notes Research
Molecular Formula C19H14Cl2N6O
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (241.98 mM)
SMILES ClC1=C(Cl)C(N2N=NN=C2NCC3=CC=CC=C3OC4=NC=CC=C4)=CC=C1
Molecular Weight (g/mol) 413.26
Formula Weight 413.26
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.