missing translation for 'onlineSavingsMsg'
Learn More

A-867744, MedChemExpress

Product Code. 16481237
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481237 50 mg 50mg
16465756 100 mg 100mg
16444998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481237 Supplier Medchem Express Supplier No. HY12149
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress A-867744 is a highly potent and selective type II positive allosteric modulator (PAM) of the alpha7 nicotinic acetylcholine receptors (nAChR) with an EC50 of 1.0 μM.

A-867744 is a highly potent and selective type II positive allosteric modulator (PAM) of the alpha7 nicotinic acetylcholine receptors (nAChR) with an EC50 of 1.0 μM.

Specifications

Chemical Name or Material A-867744
CAS 1000279-69-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H19ClN2O3S
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (248.21 mM)
SMILES O=S(C1=CC=C(C=C1)N2C(C)=C(C=C2C3=CC=C(C=C3)Cl)C(CC)=O)(N)=O
Molecular Weight (g/mol) 402.89
Formula Weight 402.89
Percent Purity 95.06%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.