missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ A33 Recombinant Mouse Monoclonal Antibody (K92070_5D9)
GREENER_CHOICE

Product Code. 30828528
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30828528 100 μL -
30830378 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828528 Supplier Invitrogen™ Supplier No. MA564443

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: LRASQGKSVT LPCTYHTSTS SREGLIQWDK LLLTHTERVV IWPFSNKNYI HGELYKNRVS ISNNAEQSDA SITIDQLTMA DNGTYECSVS LMSDLEGNTK SRVRLLVLVP PSKPECGIEG ETIIGNNIQL TCQSKEGSPT PQYSWKRYNI LNQEQPLAQP ASGQPVSLKN ISTDTSGYYI CTSSNEEGTQ Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 33%, Pig - 71%, Cynomolgus monkey - 90%.

The glycoprotein encoded by this gene is a cell surface antigen that is expressed in greater than 95% of human colon cancers. The open reading frame encodes a 319-amino acid polypeptide having a putative secretory signal sequence and 3 potential glycosylation sites. The predicted mature protein has a 213-amino acid extracellular region, a single transmembrane domain, and a 62-amino acid intracellular tail. The sequence of the extracellular region contains 2 domains characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A33
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K92070_5D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene Gpa33
Gene Accession No. Q99795
Gene Alias 2010310L10Rik; 2210401D16Rik; A33; A33 antigen; BB116197; cell surface A33 antigen; Glycoprotein A33; glycoprotein A33 (transmembrane); GPA33; mA33; MGC129986; MGC129987; transmembrane glycoprotein A33
Gene Symbols Gpa33
Host Species Mouse
Immunogen Recombinant human GPA33 (aa 31-220).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10223
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.