missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ A33 Recombinant Mouse Monoclonal Antibody (K92070_5D9)

Code produit. 30830378
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 μL
100 μL
missing translation for 'unitSize':
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantity missing translation for 'unitSize'
30830378 50 μL 50µL
30828528 100 μL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 30830378 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' MA565902
 Restricted Item
Estimated Shipment: 22-12-2026
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: LRASQGKSVT LPCTYHTSTS SREGLIQWDK LLLTHTERVV IWPFSNKNYI HGELYKNRVS ISNNAEQSDA SITIDQLTMA DNGTYECSVS LMSDLEGNTK SRVRLLVLVP PSKPECGIEG ETIIGNNIQL TCQSKEGSPT PQYSWKRYNI LNQEQPLAQP ASGQPVSLKN ISTDTSGYYI CTSSNEEGTQ Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 33%, Pig - 71%, Cynomolgus monkey - 90%.

The glycoprotein encoded by this gene is a cell surface antigen that is expressed in greater than 95% of human colon cancers. The open reading frame encodes a 319-amino acid polypeptide having a putative secretory signal sequence and 3 potential glycosylation sites. The predicted mature protein has a 213-amino acid extracellular region, a single transmembrane domain, and a 62-amino acid intracellular tail. The sequence of the extracellular region contains 2 domains characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily.
TRUSTED_SUSTAINABILITY

Spécification

Antigen A33
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K92070_5D9
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene Gpa33
Gene Accession No. Q99795
Gene Alias 2010310L10Rik; 2210401D16Rik; A33; A33 antigen; BB116197; cell surface A33 antigen; Glycoprotein A33; glycoprotein A33 (transmembrane); GPA33; mA33; MGC129986; MGC129987; transmembrane glycoprotein A33
Gene Symbols Gpa33
Host Species Mouse
Immunogen Recombinant human GPA33 (aa 31-220).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10223
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.