Learn More
AADACL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14021-1-AP-20UL
 View more versions of this product
Product Code. 16852484
  • 145.00 € / 20µL
Description
Neutral cholesterol ester hydrolase 1 (NCEH) also known as arylacetamide deacetylase-like 1 (AADACL1) or KIAA1363 is an enzyme that in humans is encoded by the NCEH1 gene. NCEH is an enzyme located in the endoplasmic reticulum. NCEH hydrolyzes 2-acetyl monoalkylglycerol ether, as part of an enzymatic pathway regulating the levels of platelet activating factor and lysophospholipids that may be involved in cancer development.
Specifications
Specifications
AADACL1
Polyclonal
Unconjugated
NCEH1
AADACL1, KIAA1363, KIAA1363, NCEH1,NCEH, NCEH, NCEH1
Rabbit
Antigen Affinity Chromatography
RUO
294930, 320024, 57552
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
B2GV54, Q6PIU2, Q8BLF1
Nceh1
AADACL1 Fusion Protein Ag5136
20 μL
Primary
Rat, Mouse, Human