missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AADACL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550286
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total AADACL2 protein.
AADACL2 Polyclonal antibody specifically detects AADACL2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
AADACL2 | |
Polyclonal | |
Unconjugated | |
AADACL2 | |
Aadacl2; ADCL2; arylacetamide deacetylase like 2; arylacetamide deacetylase-like 2; EG639634; MGC72001; RGD1563197 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
344752 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q6P093 | |
AADACL2 | |
Fusion protein corresponding to residues near the C terminal of human arylacetamide deacetylase-like 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |