missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AAK1 Monoclonal Antibody (702425)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561733
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
AAK1 serine/threonine kinase is a member of the NAK family and plays a regulatory role in endocytosis. AAK1 is a member of the SNF1 subfamily of Ser/Thr protein kinases. The protein interacts with and phosphorylates a subunit of the AP-2 complex, which promotes binding of AP-2 to sorting signals found in membrane-bound receptors and subsequent receptor endocytosis. Its kinase activity is stimulated by clathrin.
Specifications
| AAK1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q2M2I8 | |
| AAK1 | |
| E. coli-derived recombinant human AAK1, Ser704-Ile822. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 702425 | |
| Unconjugated | |
| AAK1 | |
| 5530400K14Rik; 9630042K20; Aak1; Adaptor-associated kinase 1; AP2 associated kinase 1; AP2-associated protein kinase 1; AU067724; AU067726; BC028270; C79663; D6Ertd245e; DKFZp686F03202; DKFZp686K16132; FLJ23712; FLJ25931; FLJ31060; FLJ42882; FLJ45252; KIAA1048; MGC138170; MGC164568; MGC164570; mKIAA1048; R75501; RGD1563580 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 22848 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction