missing translation for 'onlineSavingsMsg'
Learn More

Abamectin >80%, TRC

Product Code. 30138535
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138535 10 g 10g
18041030 250 mg 250mg
18022220 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138535 Supplier Toronto Research Chemicals Supplier No. TRCA10700010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71751-41-2
Molecular Formula C48H72O14
Molecular Weight (g/mol) 873.08
Synonym Avermectin B1, Spiro[11,15-methano-2H,13H,17H-furo[4,3,2-pq][2,6]benzodioxacyclooctadecin-13,2'-[2H]pyran], avermectin B1 deriv., A 14906, A 8612, Abamectin, Abamektin, Abamitel, Abarex, Abastate, Abastate EW, Abba, AgMectin, Agri-Mek, Agrimec, Avermectin B1a-Avermectin B1b mixt., Aversectin S, Avert, Avicta, Avicta 400FS, Avid, Avid 0.15EC, CHA 2061, CHA 2062, Epi-Mek, Fitoverm, Genesis Oral Drench, Genesis Pour-on for Cattle and Deer, Kraft, L 676, Lirosect, Lyrosekt, MK 936, Mectin, PT 310, Phytoverm, Rustomectin, STAN, Temprano, Vermitec, Vertigo (pesticide), Vertigo 018EC, Vertimec, Vertimec 018SC, Zephyr, Zoro
IUPAC Name (2S,2a'E,2a1'S,4'E,5S,6R,6'S,7'S,8'E,11'R,15'S,17a'R,20'R,20a'R)-6-((S)-sec-butyl)-2a1',20'-dihydroxy-7'-(((2R,4S,5S,6S)-5-(((2S,4S,5S,6S)-5-hydroxy-4-methoxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-4-methoxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-5,6',8',19'-tetramethyl-2a1',5,6,6',7',10',11',14',15',17a',20',20a'-dodecahydro-2'H,17'H-spiro[pyran-2,13'-[11,15]methano[1,5]dioxacyclooctadecino[9,8,7-cd]benzofuran]-17'-one
SMILES CO[C@H]1C[C@H](O[C@H]2[C@H](C)O[C@H](C[C@@H]2OC)O[C@H]3[C@@H](C)\C=C\C=C\4/CO[C@@H]5[C@H](O)C(=C[C@@H](C(=O)O[C@H]6C[C@@H](C\C=C\3/C)O[C@@]7(C6)O[C@H](C8CCCCC8)[C@@H](C)C=C7)[C@]45O)C)O[C@@H](C)[C@@H]1O

Specifications

Analyte or Component Names Abamectin
Shipping Condition Room Temperature
InChI Formula InChI=1S/C48H72O14/c1-11-25(2)43-28(5)17-18-47(62-43)23-34-20-33(61-47)16-15-27(4)42(26(3)13-12-14-32-24-55-45-40(49)29(6)19-35(46(51)58-34)48(32,45)52)59-39-22-37(54-10)44(31(8)57-39)60-38-21-36(53-9)41(50)30(7)56-38/h12-15,17-19,25-26,28,30-31,33-45,49-50,52H,11,16,20-24H2,1-10H3/b13-12+,27-15+,32-14+/t25-,26-,28-,30-,31-,33+,34-,35-,36-,37-,38-,39-,40+,41-,42-,43+,44-,45+,47+,48+/m0/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 10 g
Formula Weight 1730.9688
Chemical Name or Material Abamectin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.