Learn More
ABAT Mouse anti-Human, Clone: OTI3D2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF806896
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ABAT (4-aminobutyrate aminotransferase) is responsible for catabolism of gamma-aminobutyric acid (GABA), an important, mostly inhibitory neurotransmitter in the central nervous system, into succinic semialdehyde. The active enzyme is a homodimer of 50-kD subunits complexed to pyridoxal-5-phosphate. The protein sequence is over 95% similar to the pig protein. GABA is estimated to be present in nearly one-third of human synapses. ABAT in liver and brain is controlled by 2 codomit alleles with a frequency in a Caucasian population of 0.56 and 0.44. The ABAT deficiency phenotype includes psychomotor retardation, hypotonia, hyperreflexia, lethargy, refractory seizures, and EEG abnormalities. Multiple alternatively spliced transcript variants encoding the same protein isoform have been found for this gene.Specifications
| ABAT | |
| Monoclonal | |
| Unconjugated | |
| ABAT | |
| GABA-AT, GABAT, NPD009 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 18 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P80404 | |
| ABAT | |
| Human recombit protein fragment corresponding to amino acids 29-323 of human ABAT produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.