missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 357 publications
307.00 € - 625.00 €
Specifications
| Dilution | Western Blot 1:500, Simple Western 1:50, Chromatin Immunoprecipitation reported in scientific literature (PMID 19515742), Flow Cytometry 1:400. Use reported in scientific literature (PMID 21501868), ELISA reported in scientific literature (PMID 18541924), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100. Use reported in scientific literature (PMID 21501868), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 21846716; 21106520), Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen, Immunoblotting reported in scientific literature (PMID 27599291), Gel Super Shift Assays reported in scientific literature (PMID 15684432), Gel Supershift Assay, Chromatin Immunoprecipitation (ChIP), Knockout Validated reported in scientific literature (PMID 32273567), KnockDown Validated reported in scientific literature (PMID 31666189), Block/Neutralize reported in scientific literature (PMID 30821416), PCR reported in scientific literature (PMID 27406916), Dual RNAscope ISH-IHC |
|---|---|
| Classification | Polyclonal |
| Regulatory Status | RUO |
| Target Species | Pig |
| Gene Alias | ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18700584
|
Novus Biologicals
NB400-105SS |
307.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18033854
|
Novus Biologicals
NB400-105 |
0.1 mL |
625.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABCA1 Polyclonal specifically detects ABCA1 in Human, Mouse, Rat, Porcine, Canine, Chicken, Chinese Hamster, Equine, Hamster, Mustelid, Primate, Rabbit samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Gel Supershift Assay, Knockout Validated, Knockdown Validated, Block/Neutralize, PCR, Dual RNAscope ISH-IHC.Specifications
| Western Blot 1:500, Simple Western 1:50, Chromatin Immunoprecipitation reported in scientific literature (PMID 19515742), Flow Cytometry 1:400. Use reported in scientific literature (PMID 21501868), ELISA reported in scientific literature (PMID 18541924), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100. Use reported in scientific literature (PMID 21501868), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 21846716; 21106520), Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen, Immunoblotting reported in scientific literature (PMID 27599291), Gel Super Shift Assays reported in scientific literature (PMID 15684432), Gel Supershift Assay, Chromatin Immunoprecipitation (ChIP), Knockout Validated reported in scientific literature (PMID 32273567), KnockDown Validated reported in scientific literature (PMID 31666189), Block/Neutralize reported in scientific literature (PMID 30821416), PCR reported in scientific literature (PMID 27406916), Dual RNAscope ISH-IHC | |
| RUO | |
| ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1 | |
| IgG |
| Polyclonal | |
| Pig | |
| Partial peptide sequence (within residues 1100-1300) of human ABCA1 [UniProt# O95477]. Actual immunogen sequence is proprietary information. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title