missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Host Species: Rabbit
- Clonality: Monoclonal
- Isotype: IgG
- Applications: Flow Cyt (Intra), ICC/IF, IP, WB
- Species Reactivity: Human, Mouse and Rat
- Storage Buffer: pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
- Form: Liquid
- Purification: Affinity purification Protein A
Specifications
Specifications
| Antigen | COL3A1 |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | EPR17673 |
| Conjugate | Unconjugated |
| Formulation | pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA |
| Gene | COL3A1 |
| Gene Symbols | COL3A1 |
| Host Species | Rabbit |
| Purification Method | Affinity purification Protein A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?