missing translation for 'onlineSavingsMsg'
Learn More

Abcam Compound 6877002, CD40-TRAF6 interaction inhibitor, 10 mg

Product Code. 32118201 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32118201 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32118201 Supplier Abcam Supplier No. ab14682910mg
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody, with 1 citations.

  • Storage Buffer: Constituents: 100% (E)-3-(2,5-dimethylanilino)-1-phenylprop-2-en-1-one
  • Form: Solid

Specifications

Conjugate Unconjugated
Formulation Constituents: 100% (E)-3-(2,5-dimethylanilino)-1-phenylprop-2-en-1-one
Gene Symbols CD40-TRAF6
Quantity 10 mg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Target Species Non-species specific
Product Type Secondary Antibody
Shipping Condition Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.