missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14628-1-AP-150UL
This item is not returnable.
View return policy
Description
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The function of this mitochondrial protein is unknown.Specifications
| ABCB10 | |
| Polyclonal | |
| Unconjugated | |
| ABCB10 | |
| ABC transporter 10 protein, ABCB10, EST20237, M ABC2, MTABC2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23456, 361439, 56199 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9JI39, Q9NRK6 | |
| Abcb10 | |
| ABCB10 Fusion Protein Ag6203 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction