missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
208.00 € - 484.00 €
Specifications
| Antigen | ABCB5 |
|---|---|
| Concentration | 1.11 mg/mL |
| Dilution | Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18273827
|
Novus Biologicals
NBP1-77689SS |
0.025 mL |
208.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18781704
|
Novus Biologicals
NBP1-77689 |
484.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
ABCB5 Polyclonal specifically detects ABCB5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| ABCB5 | |
| Immunocytochemistry/Immunofluorescence 1:100 | |
| Polyclonal | |
| Rabbit | |
| ABC Transporters, Lipid and Metabolism | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5 | |
| ABCB5 | |
| IgG | |
| Affinity Purified |
| 1.11 mg/mL | |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| Q2M3G0 | |
| 340273 | |
| A synthetic peptide made to an internal portion of the human ABCB5 protein (between residues 450-500) [UniProt Q2M3G0] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title