missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABCB7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15555432
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15555432 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15555432 Supplier Invitrogen™ Supplier No. PA542395

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: DEATSSLDSI TEETILGAMK DVVKHRTSIF IAHRLSTVVD ADEIIVLDQG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 85%; Zebrafish: 92%.

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. This gene encodes a half-transporter involved in the transport of heme from the mitochondria to the cytosol. With iron/sulfur cluster precursors as its substrates, this protein may play a role in metal homeostasis. Mutations in this gene have been implicated in X-linked sideroblastic anemia with ataxia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCB7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ABCB7
Gene Accession No. O75027
Gene Alias AA517758; ABC transporter 7 protein; ABC7; ABCB7; ASAT; Atm1p; ATP binding cassette subfamily B member 7; ATP-binding cassette 7; ATP-binding cassette sub-family B member 7; ATP-binding cassette sub-family B member 7, mitochondrial; ATP-binding cassette sub-family B member 7, mitochondrial-like protein; ATP-binding cassette transporter 7; ATP-binding cassette, subfamily B (MDR/TAP), member 7; ATP-binding cassette, sub-family B (MDR/TAP), member 7; ATP-binding cassette, sub-family B, member 7; ATP-binding cassette, sub-family B, member 7, mitochondrial; AU019072; EST140535; fc20c02; wu:fc20c02
Gene Symbols ABCB7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ABCB7 (aa 633-682).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.