Learn More
ABCC10 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30697-1-AP-20UL
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
The ABCC10 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ABCC10. This antibody recognizes human, mouse antigen. The ABCC10 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Tekniske data
| ABCC10 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| ABCC10, MRP7, SIMRP7 | |
| 20 μL | |
| 89845 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion