missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG1 Antibody (S06-5B9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20076-100ul
This item is not returnable.
View return policy
Description
ABCG1 Monoclonal antibody specifically detects ABCG1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ABCG1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S06-5B9 | |
| Unconjugated | |
| ABC transporter 8, ABC8, ABCG1, ATP-binding cassette sub-family G member 1, ATP-binding cassette transporter 8, ATP-binding cassette transporter member 1 of subfamily G, ATP-binding cassette, sub-family G (WHITE), member 1, homolog of Drosophila white, MGC34313, White protein homolog, white protein homolog (ATP-binding cassette transporter 8), WHITE1, WHT1 | |
| A synthetic peptide of human ABCG1 (Uniprot # P45844) | |
| 100 μg | |
| ABC Transporters, Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Neuroscience, Signal Transduction | |
| 9619 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction