missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
386.00 € - 529.00 €
Specifications
| Antigen | ABCG8 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18439190
|
Novus Biologicals
NBP1-83388-25ul |
25 μL |
386.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18244436
|
Novus Biologicals
NBP1-83388 |
0.1 mL |
529.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABCG8 Polyclonal specifically detects ABCG8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ABCG8 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q9H221 | |
| 64241 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:HMVQYFTAIGYPCPRYSNPADFYVDLTSIDRRSREQELATREKAQSLAALFLEKVRDLDDFLWKAETKDLDEDTCVESSVTPLDTNCLPSPTKMPGAVQQFTTLIRRQISNDFRDLP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 76 kDa |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| ABC Transporters, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ATP-binding cassette sub-family G member 8, ATP-binding cassette, sub-family G (WHITE), member 8, ATP-binding cassette, sub-family G (WHITE), member 8 (sterolin 2), GBD4ATP-binding cassette, subfamily G, member 8, MGC142217, sterolin 2, sterolin-2, STSL | |
| ABCG8 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title