missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PCR Plate, 384-well, standard, white

Product Code. 10273513
Change view
Click to view available options
Quantity:
50 plates
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10273513 50 plates Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10273513 Supplier Thermo Scientific™ Supplier No. AB1384W
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ ABgene 384-Well PCR Plate Full Skirted PCR Plates are thin walled for efficient heat transfer and are compatible with most major thermal cyclers.

Thermo Scientific™ ABgene™ 384-Well Full Skirted PCR Plates are thin walled for efficient heat transfer and compatible with most major thermal cyclers.

  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer for equally high performance from every sample
  • SBS footprint
  • 40μL maximum well volume, 25μL working volume
  • Cut Corner: A24

Industry-leading Manufacturing and QC Testing for ultimate product reliability:

  • Cleanroom Produced—To avoid particulate and biological contaminants that can interfere with molecular biology applications
  • Virgin Medical Grade Polypropylene—Specialized polymer is highly inert and will not interfere with PCR reaction components
  • Integrity Testing—Every well of every plate is visually inspected and tested using an electrostatic pinhole detection method
  • PCR Functional Assays—Products are tested in PCR assays to verify clean negative controls and consistent technical replicates
  • Evaporation Testing—Samples from each lot are run through PCR cycling to test sealing performance
  • Biological Testing—All lots are biologically tested to certify them free of RNase, DNase, and DNA

Format Options:

Bar Coded Plates

  • Feature two industry standard 128 bar codes carefully designed and positioned for compatibility with all major bar code readers
  • Codes are random and each bar code also includes a human readable format
  • Wide temperature tolerance (-196° to +120°C)
  • Proprietary coating for superior scratch-resistance
  • Precise sizing and placement for reliable scanning
  • Custom bar coding also available, please enquire

White Plates

  • Designed specifically for qPCR

  • Reflect significantly more signal than traditional clear plates, which ensures that even the lowest levels of fluorescence are detected
  • Enable consistent signal reflection—the opaque well walls prevent signal from passing through to the cycler block where it can be inconsistently reflected or absorbed, keeping variations in the cycler block from affecting your qPCR data

Colored Plates

  • Great for sample organization and tracking
  • Available in five distinct colors: Blue, Green, Purple, Red, and Yellow

Compatible with:

Sealing Options: Adhesive Sealing, Heat Sealing.

Recommended for:

PCR; qPCR / Real-Time PCR

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Certified RNase/DNase-free
Packaging 50 Plate
Color White
Product Type PCR Microplate
For Use With (Equipment) Real-Time PCR Systems or Thermal Cyclers
Skirt Style Skirted
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 25 μL
Quantity 50 plates
Reaction Speed Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.