missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
GREENER_CHOICE

Product Code. 10602674
Change view
Click to view available options
Color:
Black
Natural
Volume (Metric) Well:
55 μL
120 μL
250 μL
300 μL
Well Shape:
Round, Conical Bottom (V-well)
Square, Conical Bottom (V-well)
Fit for Purpose:
PCR Ready
Unit Size:
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Volume (Metric) Well Well Shape Fit for Purpose unitSize
10602674 Natural 250 μL Square, Conical Bottom (V-well) PCR Ready Pack of 50
10701534 Natural 120 μL Square, Conical Bottom (V-well) PCR Ready Pack of 50
10324663 Black 120 μL Square, Conical Bottom (V-well) PCR Ready Pack of 50
11514764 Natural 300 μL Square, Conical Bottom (V-well) PCR Ready Pack of 50
10532255 Natural 55 μL Round, Conical Bottom (V-well) PCR Ready Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10602674 Supplier Thermo Scientific™ Supplier No. AB1178
Only null left
Add to basket
Add to basket

384-well microarray plate with pyramidal well bottoms for maximum sample recovery.

Thermo Scientific™ Abgene™ polypropylene microplates and DeepWell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use and have molded slots and pads for automated sample processing and preparation workflows. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using USP <661.1> high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and DeepWell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.

All Thermo Scientific plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. The Virgin medical grade resin passes requirements for USP <661.1> and USP <665> reports have been completed for both CapMat sealed and Heat Sealed plates.

 

Highlights

  • Certified free from DNAse, RNAse, PCR Inhibitors, and human DNA
  • Pyramidal well bottom for ease of sample retrieval
  • Discrete raised rims around each well for effective sealing and to reduce the risk of cross-contamination

Compatible Sealing Film: Adhesive and Heat
Hazardous: No
Max Centrifugal Force: 2000-5000 RCF
Storage Condition: Ambient stored, or transported

All plates are ANSI format for compatibility with automated systems.

To further assist in sample storage and tracking, all plates can be customized with non-repeating barcoding and labelling. OEM options for custom logos, colors and full packaging support are available.

TRUSTED_SUSTAINABILITY

Specifications

Barcode Custom Barcoding Available
Certifications/Compliance Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Format Standard
Description 384-well Polypropylene Sample Processing & Storage Plates
Array 16 x 24
Autoclavable Autoclavable
Packaging 10 pk., 50 cs.
Binding Type Low binding PP
Product Line Abgene
Color Natural
Product Type 384-well plate
For Use With (Equipment) ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
Material Polypropylene
Volume (Metric) Well 250 μL
No. of Wells 384
Volume (Metric) Working 200 μL
Sterility Cleanroom
Surface Treatment Non-treated
Well Shape Square, Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
Max. RCF 5000 x g
No. per Case 50
No. per Pack 10
Regulatory Status For Research Use Only. Not for use in diagnostic procedures.
RNase Free Yes
Fit for Purpose PCR Ready
Plate Size ANSI/ SLAS Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.