missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates

Product Code. 11554754
Change view
Click to view available options
Well Shape:
Round
Round, Conical Bottom (V-well)
Volume (Metric) Well:
200 μL
330 μL
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Shape Volume (Metric) Well Unit Size
11554754 Round 330 μL Pack of 100
10304513 Round, Conical Bottom (V-well) 200 μL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11554754 Supplier Thermo Scientific™ Supplier No. AB0796
  • ONLINE EXCLUSIVE PRICE
    244.23 €
    191.01 € / Pack of 100
    Save 53.22 €
    21% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

Streamline your molecular biology applications with these 96-well Polypropylene Storage Plates.

Thermo Scientific Abgene 96 Well Polypropylene Storage Microplates provide excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. All plates are ANSI format for compatibility with automated systems and are manufactured in our cleanroom facility to ensure molecular grade quality. To further assist in storage and tracking, all plates can be supplied with custom barcoding.

Features

  • Alphanumeric grid-referencing and industry standard footprint
  • Polypropylene
  • Sealing options: Adhesive Seals, Heat Seals and Cap Strips
  • Cleanroom manufactured

Applications

  • Sample storage
  • Sample resuspension
  • Cell culture
  • Assay setup: diluting and aliquoting
TRUSTED_SUSTAINABILITY

Specifications

Barcode Custom Barcoding Available
Certifications/Compliance Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Format Standard
Description 96-well Polypropylene Sample Processing & Storage Plates
Array 8 x 12
Autoclavable Autoclavable
Packaging Bulk
Binding Type Low binding PP
Product Line Abgene
Color Natural
Product Type 96-well plate
For Use With (Equipment) ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
Material Polypropylene
Volume (Metric) Well 330 μL
No. of Wells 96
Volume (Metric) Working 300 μL
Sterility Sterile
Surface Treatment Non-treated
Well Shape Round
Dnase Free Yes
For Use With (Application) Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
Max. RCF 5000 x g
No. per Case 100
No. per Pack 1
Regulatory Status RUO
RNase Free Yes
Fit for Purpose PCR Ready
Plate Size ANSI/ SLAS Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.