missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABI3 Recombinant Rabbit Monoclonal Antibody (3V2G1)

Rabbit Recombinant Monoclonal Antibody
176.00 € - 436.00 €
Specifications
| Antigen | ABI3 |
|---|---|
| Clone | 3V2G1 |
| Concentration | 1.08 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30283053
|
Invitrogen™
MA558083 |
20 μL |
176.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30282476
![]() |
Invitrogen™
MA558084 |
100 μL |
436.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: VPDGRLSAAS SAFSLASAGS AEGVGGAPTP KGQAAPPAPP LPSSLDPPPP PAAVEVFQRP PTLEELSPPP PDEELPLPLD LPPPPPLDGD ELGLPPPPPG FGPDEPSWVP ASYLEKVVTL YPYTSQKDNE LSFSEGTVIC VTRRYSDGWC EGVSSEGTGF FPGNYVEPSC.
Inhibits ectopic tumor cell metastasis of SRD cells. In vitro, reduces cell motility.Specifications
| ABI3 | |
| 1.08 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9P2A4 | |
| 51225 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 197-366 of human ABI3 (NP_057512.2). | |
| Antibody |
| 3V2G1 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| ABI3 | |
| 2210414K06Rik; ABI family member 3; ABI family, member 3; ABI gene family member 3; ABI gene family, member 3; Abi3; AI987680; NESH; NESH protein; new molecule including SH3; SSH3BP3 | |
| ABI3 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title