missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABLIM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | ABLIM1 |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600887
|
Novus Biologicals
NBP2-99386-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605546
|
Novus Biologicals
NBP2-99386-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABLIM1 Polyclonal antibody specifically detects ABLIM1 in Human samples. It is validated for Western BlotSpecifications
| ABLIM1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ABLIM, abLIM-1, actin binding LIM protein 1, Actin-binding double zinc finger protein, actin-binding double-zinc-finger protein, actin-binding LIM protein 1, Actin-binding LIM protein family member 1, DKFZp781D0148, FLJ14564, KIAA0059, LIM actin-binding protein 1, LIMAB1MGC1224, LIMATIN | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ABLIM1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 3983 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title